Order Entry
Northern Ireland
ContactUsLinkComponent
Tris(2-methylphenyl)phosphine 99%
Irritant
Tris(2-methylphenyl)phosphine 99%
Certificates
undefined
Irritant
Tris(2-methylphenyl)phosphine 99%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    122 to 126 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together