Tetraoctylammonium bromide, Supelco®
Catalog # 87996-10G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:546.8 g/mol
Melting Pt:~ 96 °C
:M-Clarity™ quality level = MQ100
Flash Pt:100 °C
EINECS:238-936-9
CAS Number:14866-33-2
Formula:C₃₂H₆₈BrN
MDL Number:MFCD00011863