
Tetramethylammonium bromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-20-0
:
Formula:C₄H₁₂BrN
MDL Number:MFCD00011626
:154.05 g/mol
Boiling Pt:∼360 °C (1013 hPa)
Melting Pt:230 °C (dec.)
Density:1,56 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-581-2
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 %
- Water:≤1 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together