
Tetrahexylammonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4328-13-6
:
Formula:C₂₄H₅₂BrN
MDL Number:MFCD00011858
:434.59 g/mol
Melting Pt:∼ 95…100 °C
Density:1,0036 g/cm³ (25 °C)
EINECS:224-363-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together