


Tetrabutylphosphonium bromide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3115-68-2
:
Formula:C₁₆H₃₆BrP
MDL Number:MFCD00011853
:339.34 g/mol
Melting Pt:99…103 °C
Flash Pt:290 °C (554 °F)
EINECS:221-487-8
UN:3464
ADR:6.1,II
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together