Order Entry
Northern Ireland
ContactUsLinkComponent
Cetrimonium bromide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Cetrimonium bromide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Cetrimonium bromide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 1 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Related Information

Frequently Bought Together