Trioctylphosphine
Catalog # MOLE73693902-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:370.64 g/mol
Boiling Pt:150…155 °C (0,03 mmHg)
Melting Pt:48 °C
Density:0,831 g/cm³ (20 °C)
EINECS:225-234-2
UN:1760
ADR:8,II
CAS Number:4731-53-7
Formula:C₂₄H₅₁P
MDL Number:MFCD00015298