Order Entry
Northern Ireland
ContactUsLinkComponent
Trioctylphosphine oxide for synthesis, Sigma-Aldrich®
 
 
Trioctylphosphine oxide for synthesis, Sigma-Aldrich®
  8.14868.4000
 :  Merck
 :  
 
 
Trioctylphosphine oxide for synthesis, Sigma-Aldrich®
  8.14868.4000
 :  Merck
 :  

 

  • Pk:
    4 kg
  • Seq:
    4102

 

 

  • Solubility:
     - 1 g/l (20 °C)
  • Melting Point:
    50 - 54 °C
  • Molar Mass:
    386.64 g/mol
  • Boiling Point:
    201 - 202 °C (3.60 hPa)
  • Flash Point:
    252 °C
  • Density:
    0.88 g/cm3 (25 °C)
  • Ignition Point:
    320 °C
  • Hill Formula:
    C24H51OP
  • Chemical Formula:
    (C8H17)3PO

 

 

 :
 :386.64 g/mol
 :409 °C (1013 hPa)
 :50…52 °C
 :0,88 g/cm³ (20 °C)
 :Ambient
 :201-121-3
 :3077
 :9,III
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
 :78-50-2
 :C₂₄H₅₁OP
 :MFCD00002083