
(1-Pentyl)triphenylphosphonium bromide
Catalog # MOLE19517443-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:413.34 g/mol
Melting Pt:168 °C
EINECS:244-374-5
:GHS07
CAS Number:21406-61-1
Formula:H₃C(CH₂)₄P(C₆H₅)₃Br
MDL Number:MFCD00031630