

Trimethyl phosphate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:512-56-1
:
Formula:C₃H₉O₄P
MDL Number:MFCD00008348
:140.08 g/mol
Boiling Pt:197,2 °C (1013 hPa)
Melting Pt:–46 °C
Density:1,21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>150 °C
Storage Temperature:Ambient
EINECS:208-144-8
:GHS08
:GHS07
Specification Test Results
- Solubility:1000 g/l (25 °C)
- Melting Point:-46 °C
- Molar Mass:140.08 g/mol
- Boiling Point:197 °C (1013 hPa)
- Vapor Pressure:1.133 hPa (25 °C)
- Flash Point:150 °C
- Refractive Index:1.3967 (20 °C)
- Density:1.21 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together