Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Triethyl phosphate ≥98%

Triethyl phosphate ≥98%

Irritant
Triethyl phosphate ≥98%
Certificates
Triethyl phosphate ≥98%
Triethyl phosphate ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 78-40-0 Molecular Formula: (CH3CH2O)3P(O) Formula Weight: 182.16 Flash Point: 115°(240°F) Refractive Index: 1.4050 Odour: Mild Boiling Point: 215° Merck Reference: 14,9674 MDL No.: MFCD00009077

  • CAS Number:78-40-0
  • Formula:C₆H₁₅O₄P
  • MDL Number:MFCD00009077
  • Molecular Weight:182.16 g/mol
  • Merck Index:14,9674
  • Boiling Pt:215 °C
  • Melting Pt:–57 °C
  • Density:1,072
  • Flash Pt:115 °C (240 °F)
  • Storage Temperature:Ambient
  • EINECS:201-114-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £45.60
    £45.60
    1 * 500 g
       £45.60

  • Glass bottle for solids
    2.500 g
    Log in for availability
    Log in for availability
    1 * 2.500 g
    1 * 2.500 g - £164.00
    £164.00
    1 * 2.500 g
       £164.00