Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium dihydrogen phosphate ≥99.0% (by acidimetry) ACS analytical reagent
Potassium dihydrogen phosphate ≥99.0% (by acidimetry) ACS analytical reagent
Supplier:  PanReac AppliChem
Certificates
Potassium dihydrogen phosphate ≥99.0% (by acidimetry) ACS analytical reagent
Supplier:  PanReac AppliChem

Specification Test Results

  • Minimum Assay (Acidim.):
    99.0%
  • pH of 5% Solution:
    4.2 - 4.5
  • Insoluble Matter in H₂O:
    Max. 0.01%
  • Loss on Drying at 105 °C:
    Max. 0.2%
  • Chloride (Cl):
    Max. 0.0005%
  • Nitrogen Compounds (as N):
    Max. 0.001%
  • Sulphate (SO₄):
    Max. 0.003%
  • As:
    Max. 0.0002%
  • Heavy Metals (ICP-OES):
    Max. 0.001%
  • Ag (by ICP):
    5 mg/kg (ppm)
  • Al (by ICP):
    5 mg/kg (ppm)
  • Au (by ICP):
    5 mg/kg (ppm)
  • B (by ICP):
    5 mg/kg (ppm)
  • Ba (by ICP):
    5 mg/kg (ppm)
  • Be (by ICP):
    5 mg/kg (ppm)
  • Bi (by ICP):
    5 mg/kg (ppm)
  • Ca (by ICP):
    50 mg/kg (ppm)
  • Cd (by ICP):
    5 mg/kg (ppm)
  • Co (by ICP):
    5 mg/kg (ppm)
  • Cr (by ICP):
    5 mg/kg (ppm)
  • Cu (by ICP):
    5 mg/kg (ppm)
  • Fe (by ICP):
    10 mg/kg (ppm)
  • Ga (by ICP):
    5 mg/kg (ppm)
  • Ge (by ICP):
    5 mg/kg (ppm)
  • In (by ICP):
    5 mg/kg (ppm)
  • Mg (by ICP):
    5 mg/kg (ppm)
  • Mn (by ICP):
    5 mg/kg (ppm)
  • Mo (by ICP):
    5 mg/kg (ppm)
  • Na (by ICP):
    50 mg/kg (ppm)
  • Ni (by ICP):
    5 mg/kg (ppm)
  • Pb (by ICP):
    5 mg/kg (ppm)
  • Sb (by ICP):
    5 mg/kg (ppm)
  • Se (by ICP):
    5 mg/kg (ppm)
  • Si (by ICP):
    5 mg/kg (ppm)
  • Sr (by ICP):
    5 mg/kg (ppm)
  • Ti (by ICP):
    5 mg/kg (ppm)
  • Tl (by ICP):
    5 mg/kg (ppm)
  • V (by ICP):
    5 mg/kg (ppm)
  • Zn (by ICP):
    5 mg/kg (ppm)
  • Zr (by ICP):
    5 mg/kg (ppm)

Specification Test Results

Frequently Bought Together