

Bismuth(III) nitrate pentahydrate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10035-06-0
:
Formula:Bi(NO₃)₃·5H₂O
MDL Number:MFCD00149157
Boiling Pt:80 °C (760 mmHg)
Melting Pt:30 °C
Density:2,736 g/cm³ (20 °C)
EINECS:233-791-8
UN:1477
ADR:5.1,II
:485.07 g/mol
:GHS07
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystals
- Total trace metal impurities:≤0.001 %
Specification Test Results
Frequently Bought Together