Order Entry
Northern Ireland
ContactUsLinkComponent
Antimony(V) fluoride for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
Antimony(V) fluoride for synthesis, Sigma-Aldrich®
Catalog # 8.12034.0100
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Environment
Antimony(V) fluoride for synthesis, Sigma-Aldrich®
Catalog # 8.12034.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Hill Formula:
    F₅Sb
  • Chemical formula:
    SbF₅
  • Molar Mass:
    216.74 g/mol
  • Boiling point:
    149 - 150 °C (1013 hPa)
  • Density:
    2.99 g/cm³ (20 °C)
  • Melting Point:
    7 °C
  • Solubility:
    (20 °C) (rigorous decomposition)
  • Storage class:
    8B Non-combustible, corrosive hazardous materials
  • WGK:
    WGK 2 water endangering
  • R Phrase:
    R 14-20/22-34-51/53
  • S Phrase:
    S 22-26-36/37/39-45-61

Specification Test Results

About this item

 :
 :216.75 g/mol
Merck Index:13,00702
Boiling Pt:149,4 °C (1013 hPa)
Melting Pt:7 °C
Density:2,99 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-021-8
UN:1732
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS07
 :GHS09
CAS Number:7783-70-2
Formula:F₅Sb
MDL Number:MFCD00011219