Order Entry
Northern Ireland
ContactUsLinkComponent
 
Hexamethyldisilane for synthesis, Sigma-Aldrich®
 :  Merck
 
Hexamethyldisilane for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    13 - 15 °C
  • Molar Mass:
    146.37 g/mol
  • Boiling Point:
    110 - 112 °C
  • Flash Point:
    -6 °C
  • Refractive Index:
    1.4207 (20 °C, 589 nm)
  • Density:
    0.73 g/cm3 (20 °C)

 

Frequently Bought Together