


Bis(tributyltin) oxide
Catalog # MOLEM49409483
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:596.11 g/mol
Boiling Pt:475 °C (1013 hPa)
Density:1,17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-268-0
UN:2788
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:56-35-9
Formula:C₂₄H₅₄OSn₂
MDL Number:MFCD00009418