Order Entry
Northern Ireland
ContactUsLinkComponent
Tributyltin hydride 97%
Toxic
Health
Environment
Tributyltin hydride 97%
Certificates
undefined
Toxic
Health
Environment
Tributyltin hydride 97%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4720 to 1.4740 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together