


Chlorotrimethylsilane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-77-4
:
Formula:C₃H₉ClSi
MDL Number:MFCD00000502
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–58 °C
Density:0,85 g/cm³
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:200-900-5
ADR:3,II
:108.64 g/mol
UN:1298
REACH:01-2119457596-25
:GHS06
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3870 to 1.3890 (20°C, 589 nm)
- GC:≥97.5 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
Frequently Bought Together