

Bromotrimethylsilane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2857-97-8
:
Formula:C₃H₉BrSi
MDL Number:MFCD00000048
:153.09 g/mol
Boiling Pt:79 °C (760 mmHg)
Melting Pt:–43 °C
Density:1,16 g/cm³
Flash Pt:32 °C
Storage Temperature:Ambient
EINECS:220-672-0
UN:2920
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4200 to 1.4250 (20°C, 589 nm)
- Titration Argentometric:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together