Dibutyltin oxide
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 818-08-6 Molecular Formula: C8H18OSn Formula Weight: 248.92 Physical Form: Powder Density: 1.58 Appearance: White Odour: Odourless Melting Point: 300° Beilstein Reference: 4126243 UN No.: 3146 MDL No.: MFCD00001992
Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide
- CAS Number:818-08-6
- Formula:C₈H₁₈OSn
- MDL Number:MFCD00001992
- Molecular Weight:248.94 g/mol
- Melting Pt:>300 °C
- Density:1,58
- Storage Temperature:Ambient
- EINECS:212-449-1
- UN:3146
- ADR:6.1,II
2 Options Available Below
Product Details & Documents
CAS No.: 818-08-6 Molecular Formula: C8H18OSn Formula Weight: 248.92 Physical Form: Powder Density: 1.58 Appearance: White Odour: Odourless Melting Point: 300° Beilstein Reference: 4126243 UN No.: 3146 MDL No.: MFCD00001992
Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide
- CAS Number:818-08-6
- Formula:C₈H₁₈OSn
- MDL Number:MFCD00001992
- Molecular Weight:248.94 g/mol
- Melting Pt:>300 °C
- Density:1,58
- Storage Temperature:Ambient
- EINECS:212-449-1
- UN:3146
- ADR:6.1,II
Documents
Specifications for Product Family
Specifications
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



