Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dioctyltin dichloride
Dioctyltin dichloride
Dioctyltin dichloride
PROODE-MS-03444
:
LGC Standards PROMOCHEM
:
Dioctyltin dichloride
PROODE-MS-03444
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
300 mg
Seq:
2625
:
:
416.06 g/mol
:
48 °C
:
222-583-2
:
3261
:
8,III
:
GHS06
:
GHS08
:
3542-36-7
:
C₁₆H₃₄Cl₂Sn
:
MFCD00018091