Order Entry
Northern Ireland
ContactUsLinkComponent
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%
Corrosive
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%
Certificates
undefined
Corrosive
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4425 to 1.4445 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together