Order Entry
Northern Ireland
ContactUsLinkComponent
Carbon disulphide ≥99.9% ACS, Riedel-de Haën™
Flammable
Health
Irritant
Carbon disulphide ≥99.9% ACS, Riedel-de Haën™
Catalog # 676918-1L
CAS Number:  
undefined
Flammable
Health
Irritant
Carbon disulphide ≥99.9% ACS, Riedel-de Haën™
Catalog # 676918-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Sulfur dioxide (SO₂):
    complying
  • Hydrogen sulfide (H₂S):
    complying
  • APHA:
    Max. 10
  • Autoignition Temperature:
    212 °F
  • Vapor Density:
    2.67 (vs air)

Specification Test Results

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46 °C (1,013 hPa)
Melting Pt:-76 °C
Melting Pt:–76 °C
Density:1,264 g/cm³ (20 °C)
Flash Pt:-30 °C
Flash Pt:–30 °C
EINECS:200-843-6
UN:1131
ADR:3,I
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321