Order Entry
Northern Ireland
ContactUsLinkComponent
Tetramethoxysilane for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Tetramethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Tetramethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) Hydrolysis
  • Melting Point:
    -2 °C
  • Molar Mass:
    152.22 g/mol
  • Boiling Point:
    122 °C (1013 hPa)
  • Vapor Pressure:
    18 hPa (20 °C)
  • Flash Point:
    26 °C
  • Explosion Limit:
    0.88 - 23.8 %(V)
  • Density:
    1.03 g/cm3 (20 °C)
  • Ignition Point:
    245 °C

Specification Test Results

Related Information

Frequently Bought Together