
Silicon carbide ≥99.95% (metals basis excluding B), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41085.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:409-21-2
:
Formula:SiC
MDL Number:MFCD00049531
:40.1 g/mol
Melting Pt:2700 °C
Density:3,22 g/cm³ (25 °C)
EINECS:206-991-8
:GHS07
Storage Temperature:Ambient