Germanium(IV) bromide ≥99.999% (metals basis)
40219.09
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
:
:392.23 g/mol
:187 °C
:236-612-1
:3260
:GHS05
:Ambient
:13450-92-5
:Br₄Ge
:MFCD00016115
:26 °C
:3.13
:8 II
:Ampuled under argon