Aluminium bromide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7727-15-3
:
Formula:AlBr₃
MDL Number:MFCD00003421
:266.69 g/mol
Merck Index:13,00330
Boiling Pt:265 °C (1013 hPa)
Melting Pt:97 °C
Density:3,2 g/cm³ (20 °C)
UN:1725
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:231-779-7
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:97 °C
- Molar Mass:266.69 g/mol
- Boiling Point:250 - 270 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (81 °C)
- Density:3.2 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together