Magnesium oxide substrate 10x10x0.5mm, polished two sides 100 orientation
Catalog # 41436.KT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:40.3 g/mol
Merck Index:13,05700
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3,58 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-171-9
CAS Number:1309-48-4
Formula:MgO
MDL Number:MFCD00011109