Magnesium oxide 98%, powder Ø 99% <150 µm (-100 mesh), extra pure
: Magnesia usta
:1309-48-4
:
:MgO
:MFCD00011109
:3600 °C (760 mmHg)
:2800 °C
:3.58 g/cm³
:40.3 g/mol
:13,05700
:Ambient
:215-171-9
- Environmentally Preferable:
- Appearance:White Powder
- Titration Complexometric:≥97.5 %
- Sulfate (SO4):≤2500 ppm
- Calcium oxide (CaO):≤3000 ppm
- Silicon dioxide (SiO2):≤500 ppm
- Loss on ignition:≤2 %
- Aluminium oxide (Al2O3):≤400 ppm
- Iron(III)oxide (Fe2O3):≤400 ppm
Frequently Bought Together