Magnesium oxide 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1309-48-4
:
Formula:MgO
MDL Number:MFCD00011109
:40.3 g/mol
Merck Index:13,05700
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3,58 g/cm³
EINECS:215-171-9
Specification Test Results
- Appearance:White to grey Crystalline powder
- Titration Complexometric:≥97.5 %
Specification Test Results
Frequently Bought Together