

Strontium iodide, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10476-86-5
:
Formula:I₂Sr
MDL Number:MFCD00049557
:341.43 g/mol
Merck Index:14,8844
Melting Pt:515 °C
Density:4,550
Storage Temperature:Ambient
EINECS:233-972-1
UN:3260
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together