Order Entry
Northern Ireland
ContactUsLinkComponent
Calcium carbonate 98.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent low in alkali, precipitated
Calcium carbonate 98.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent low in alkali, precipitated
Supplier:  VWR Chemicals
Certificates
undefined
Calcium carbonate 98.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent low in alkali, precipitated
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calc. on dried substance):
    98.5 - 100.5 %
  • Ba (Barium):
    Passes test
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Particle size d50 (about 14 µm):
    Passes test
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Insolubility in acetic acid:
    Max. 0.2 %
  • Insolubility in hydrochloric acid:
    Max. 50 ppm
  • Loss on drying (200°C):
    Max. 1.0 %
  • Magnesium and alkali metals:
    Max. 1.5 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 0.03 %
  • Al (Aluminium):
    Max. 50 ppm
  • As (Arsenic):
    Max. 4 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mg (Magnesium):
    Max. 0.02 %
  • Na (Sodium):
    Max. 0.2 %
  • Pb (Lead):
    Max. 5 ppm
  • Sr (Strontium):
    Max. 0.1 %
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together