Calcium carbonate 98+%, heavy powder, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C (dec.)
Density:2,71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
Specification Test Results
- Appearance:White to off-white Powder
- Titration Complexometric:≥98.0 %
- Titration with HClO4:0.40 to 0.60 M
- NMR:≥94 %
Specification Test Results
Frequently Bought Together