


Potassium peroxodisulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7727-21-1
:
Formula:K₂S₂O₈
MDL Number:MFCD00011386
:270.32 g/mol
Merck Index:13,07741
Melting Pt:100 °C
Density:2,477 g/cm³ (20 °C)
EINECS:231-781-8
UN:1492
ADR:5.1,III
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:min. 99.0 %
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 5 ppm
- Manganese (Mn):max. 2 ppm
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble Matter:max. 0.005 %
- Nitrogen Compounds (as N):max. 0.05 %
Specification Test Results
Frequently Bought Together