Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium peroxodisulphate 99+% for analysis
Oxidizing
Health
Irritant
Potassium peroxodisulphate 99+% for analysis
Certificates
undefined
Oxidizing
Health
Irritant
Potassium peroxodisulphate 99+% for analysis
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Iodometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤120 ppm
  • Iron (Fe):
    ≤20 ppm
  • Manganese (Mn):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together