Potassium hexachloroplatinate(IV) ≥99%, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Dipotassium hexachloroplatinate
CAS Number:16921-30-5
:
Formula:K₂PtCl₆
MDL Number:MFCD00011389
Density:3.344 g/cm³ (20 °C)
Molecular Weight:485.99 g/mol
Melting Pt:250
ADR:8 (6.1)
Merck Index:13,07720
:M-Clarity™ quality level = MQ100
EINECS:240-979-3
UN:2923
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity (elemental analysis):passes test
- Assay (calculated of Pt):99.0 - 101.0 %
- Pt (Platinum):39.7 - 40.5 %
- Al (Aluminium):≤ 10 ppm
- Ca (Calcium):≤ 50 ppm
- Cd (Cadmium):≤ 10 ppm
- Co (Cobalt):≤ 10 ppm
- Cr (Chromium):≤ 10 ppm
- Cu (Copper):≤ 50 ppm
- Fe (Iron):≤ 50 ppm
- Hg (Mercury):≤ 100 ppm
- Ir (Iridium):≤ 100 ppm
- Mg (Magnesium):≤ 50 ppm
- Mn (Manganese):≤ 10 ppm
- Ni (Nickel):≤ 10 ppm
- Os (Osmium):≤ 100 ppm
- Pb (Lead):≤ 50 ppm
- Pd (Palladium):≤ 100 ppm
- Rh (Rhodium):≤ 100 ppm
- Ru (Ruthenium):≤ 100 ppm
- Zn (Zinc):≤ 10 ppm
Specification Test Results
Frequently Bought Together