

Cesium chromate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13454-78-9
:
Formula:CrCs₂O₄
MDL Number:MFCD00016050
ADR:5.1,III
:381.8 g/mol
EINECS:236-640-4
UN:3087
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together