


Cesium dichromate ≥99.8% (metals basis)
Catalog # 40230.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:481.8 g/mol
Storage Temperature:Ambient
EINECS:236-879-4
UN:3087
ADR:5.1,III
:GHS03
:GHS08
:GHS07
CAS Number:13530-67-1
Formula:Cr₂Cs₂O₇
MDL Number:MFCD00049436