
Rubidium carbonate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:584-09-8
:
Formula:CO₃Rb₂
MDL Number:MFCD00011188
:230.94 g/mol
Melting Pt:837 °C
Density:2,43 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:209-530-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:4500 g/l (20 °C)
- Melting Point:837 °C
- Molar Mass:230.95 g/mol
- pH:11.7 (50 g/l, H2O, 20 °C)
- Hill Formula:CO3Rb2
- Chemical Formula:Rb2CO3
Specification Test Results
Frequently Bought Together