Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium carbonate ≥99.5%, powder ACS, Fluka™
Sodium carbonate ≥99.5%, powder ACS, Fluka™
Certificates
undefined
Sodium carbonate ≥99.5%, powder ACS, Fluka™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (excluding ignited substance):
    Min. 99.5 %
  • Loss on ignition (285°C):
    Max. 1.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.03 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Silicate (as SiO₂):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Total S (as SO₄):
    Max. 0.003 %

Specification Test Results

Related Information

Frequently Bought Together