
Cesium carbonate
Supplier: Molekula
Certificates
About this item
CAS Number:534-17-8
:
Formula:Cs₂CO₃
MDL Number:MFCD00010957
:325.82 g/mol
Merck Index:13,02020
Melting Pt:610 °C
Density:4,24 g/cm³ (20 °C)
EINECS:208-591-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together