

Cesium carbonate 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:534-17-8
:
Formula:Cs₂CO₃
MDL Number:MFCD00010957
:325.82 g/mol
Merck Index:13,02020
Melting Pt:>400 °C
Density:4,24 g/cm³ (20 °C)
EINECS:208-591-9
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Total trace metal impurities:≤0.01 %
Specification Test Results
Frequently Bought Together