

Cesium carbonate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:534-17-8
:
Formula:Cs₂CO₃
MDL Number:MFCD00010957
:325.82 g/mol
Merck Index:13,02020
Melting Pt:>400 °C
Density:4,24 g/cm³ (20 °C)
EINECS:208-591-9
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Assay:≥99.4 % (By difference)
- Loss on drying:≤0.5 % (500°C)
- Chloride (Cl):≤300 ppm
- Sulfate (SO4):≤500 ppm
- Aluminium (Al):≤100 ppm
- Barium (Ba):≤100 ppm
- Calcium (Ca):≤20 ppm
- Potassium (K):≤400 ppm
- Rubidium (Rb):≤3000 ppm
- Sodium (Na):≤300 ppm
Specification Test Results
Frequently Bought Together