Rubidium bromide ≥99.8% (metals basis)
Catalog # 12891.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:165.37 g/mol
Merck Index:14,8283
Boiling Pt:1340 °C
Melting Pt:693 °C
Density:3,35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-162-5
CAS Number:7789-39-1
Formula:BrRb
MDL Number:MFCD00011186