

Cesium bromide 99.9%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-69-1
:
Formula:BrCs
MDL Number:MFCD00010954
:212.81 g/mol
Merck Index:13,02019
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:636 °C
Density:4,44 g/cm³
EINECS:232-130-0
:GHS08
:GHS07
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Total trace metal impurities:≤0.1 %
Specification Test Results
Frequently Bought Together