


Potassium tetrahydroborate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13762-51-1
:
Formula:H₄BK
MDL Number:MFCD00011396
:53.94 g/mol
Merck Index:12,07777
Melting Pt:500 °C
Density:1,17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-360-5
ADR:4.3,I
:M-Clarity™ quality level = MQ200
UN:1870
:GHS02
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:500 °C
- Molar Mass:53.94 g/mol
- Bulk Density:400 kg/m3
- Density:1.11 g/cm3
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Specification Test Results
Frequently Bought Together