


Sodium hexafluoroaluminate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13775-53-6
:15096-52-3
Formula:AlF₆Na₃
MDL Number:MFCD00003507
:209.94 g/mol
Melting Pt:1000 °C
Density:2,90
EINECS:237-410-6
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together