Sulphuric acid 95% ≥95%, ARISTAR® for trace analysis
450061Q
:
- Pack type:Glass bottle, Safebreak®
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
- Assay:Min. 95.0 %
- Cl (Chloride):Max. 0.1 ppm
- NO3 (Nitrate):Max. 0.1 ppm
- PO4 (Phosphate):Max. 0.05 ppm
- Al (Aluminium):Max. 0.050 ppm
- As (Arsenic):Max. 0.005 ppm
- B (Boron):Max. 0.050 ppm
- Ba (Barium):Max. 0.050 ppm
- Ca (Calcium):Max. 0.100 ppm
- Cd (Cadmium):Max. 0.001 ppm
- CO:Max. 0.005 ppm
- Cu (Copper):Max. 0.010 ppm
- Fe (Iron):Max. 0.050 ppm
- Hg (Mercury):Max. 0.001 ppm
- K (Potassium):Max. 0.050 ppm
- Mg (Magnesium):Max. 0.030 ppm
- Mn (Manganese):Max. 0.005 ppm
- Na (Sodium):Max. 0.300 ppm
- NH4 (Ammonium):Max. 1.0 ppm
- Ni (Nickel):Max. 0.005 ppm
- Pb (Lead):Max. 0.005 ppm
- Se (Selenium):Max. 0.100 ppm
- Sr (Strontium):Max. 0.005 ppm
- Zn (Zinc):Max. 0.020 ppm
- Oxygen absorbing components:Max. 1 ppm
- Non-volatile matter:Max. 10 ppm
:
:98.08 g/mol
:10.38 °C
:231-639-5
:1830
:01-2119458838-20
:GHS05
:Ambient
:7664-93-9
:H₂SO₄
:MFCD00064589
:330 °C (1013 hPa)
:1.84 g/cm³ (20 °C)
:8 II