Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
Molybdic acid, Supelco®
  1.00400.1000
 :  Merck
 :  
 
 
Molybdic acid, Supelco®
  1.00400.1000
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    300 °C
  • Molar Mass:
    161.95 g/mol
  • Bulk Density:
    980 kg/m3

 

 

 :
 :13,06262
 :∼300 °C (1013 hPa)
 :300 °C
 :3,1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :161.95 g/mol
 :231-970-5
 :GHS08
 :GHS07
 :Ambient
 :7782-91-4
 :H₂MoO₄
 :MFCD00036287