Fluorescein disodium salt 98.5-100.5% indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:518-47-8
:
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039
:376.28 g/mol
Merck Index:13,04185
Melting Pt:300 °C
Density:1,601 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-253-0
Specification Test Results
- Assay (calculated to the dried substance):Min. 98.5 %
- Loss on drying (105°C):Max. 10 %
- Zinc (Zn):complying
- Chloride (Cl):Max. 0.35 %
- Sulfate (SO₄):Max. 1.2 %
- UV spectrum:complying
Specification Test Results
Frequently Bought Together